Anti-LRP1 Mouse Monoclonal Antibody

Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-863EA 3590 DKK
PRSI49-863
Anti-LRP1 Mouse Monoclonal Antibody
Antistoffer
Suitable for use in Flow Cytometry, Immunohistochemistry, ELISA, Immunoprecipitation and Western Blot. Other applications have not been tested. Recognizes human CD91, a type I membrane protein which is also known as the alpha 2 macroglobulin receptor (alpha 2 MR). Recognizes an epitope in the extracellular region of the 500kD alpha chain.

The low-density lipoprotein receptor related protein 1 (LRP1) is a cell surface protein highly expressed in liver that has close structural and biochemical similarities to the LDL receptor. LRP1 strongly binds calcium and mediates the clearance of chylomicron remnants from the plasma. LPR1 was also separately cloned as the alpha-2-macroglobulin receptor (A2MR) and is necessary for the A2M mediated clearance of beta-amyloid 40 and 42 via receptor-mediated endocytosis. LPR1 may undergo cleavage and the small C-terminal intracellular domain may be found in the cytoplasm or the nucleus of cells.

LRP1 antibody can be used in ELISA, and immunohistochemistry starting at 20 μg/mL.

Protein G Column

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LRP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR