Anti-IDH1 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-853EA 3590 DKK
PRSI49-853
Anti-IDH1 Rabbit Polyclonal Antibody
Antistoffer
IDH1 antibody can be used in immunohistochemistry starting at 2 - 4 μg/mL, and immunofluorescence.

Affinity Chromatography

PBS, pH 7.2, 0.09% sodium azide.

Store IDH1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles. Store undiluted.

Type:
Antigen: IDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR