Anti-KLK11 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-842EA 3590 DKK
PRSI49-842
Anti-KLK11 Rabbit Polyclonal Antibody
Antistoffer
Hippostasin, also known as KLK11, is a kallikrein-like serine protease, which has two alternatively spliced isoforms, brain-type and prostate-type. Hippostasin plays a role in the prostate, including reproductive and/or tumorigenic functions. Elevated serum levels of hK11 were found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of the KLK11 biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma. Suitable for use ELISA and Western Blot. Other applications not tested. Recognizes the residues 68-82 [YIVHLGQHNLQKEEG] of the 35kD human hippostasin/KLK11 protein. Species sequence homology: mouse and rat.

Kallikrein 11 antibody can be used in ELISA, and immunohistochemistry starting at 7 μg/mL.

PBS, pH 7.2, 0.02% sodium azide.

PBS, pH 7.2, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KLK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gorilla, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR