Anti-PTH1R Mouse Monoclonal Antibody

Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-809EA 3840 DKK
PRSI49-809
Anti-PTH1R Mouse Monoclonal Antibody
Antistoffer
PTHR1, a Parathyroid Hormone Receptor, mediates PTH-dependent regulation of mineral-ion homeostasis. It also mediates the paracrine action of PTHRP in endochondral bone formation. Mutations in this receptor are responsible for disorders of calcium and bone metabolism such as chondrodysplasia, hypercalcemia, and chronic renal failure. Three promoters, P1, P2, and P3, regulate the expression of PTHR.

PTHR1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 15 μg/mL.

Protein G Column

PBS, pH 7.6, 0.07% sodium azide, 30% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTH1R
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Gibbon, Bovine, Dog, Gorilla, Horse, Human, Monkey, Orangutan, Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR