Anti-HP Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-808EA 3590 DKK
PRSI49-808
Anti-HP Rabbit Polyclonal Antibody
Antistoffer
Suitable for use in ELISA. Other applications not tested. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-rabbit serum.

Protein A Column

PBS, pH 7.2, 0.01% sodium azide. No stabilizing proteins added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR