Anti-SH3GLB1 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-805EA 3590 DKK
PRSI49-805
Anti-SH3GLB1 Rabbit Polyclonal Antibody
Antistoffer
SH3GLB1 antibody can be used in immunohistochemistry starting at 20 μg/mL.

Protein A Column

PBS, pH 7.2, 50% glycerol, 1% BSA, 0.02% Thimerosal.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SH3GLB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR