Anti-CK15 Mouse Monoclonal Antibody

Leverandør: ProSci Inc.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-804EA 3590 DKK
PRSI49-804
Anti-CK15 Mouse Monoclonal Antibody
Antistoffer
KRT15 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 μg/mL, and flow cytometry. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10-20 min., followed by cooling at RT for 20 min. Immunofluorescence, Western Blotting, Immunohistochemistry Recognizes keratin 15.

Protein A Column

PBS, pH 7.4, 0.2% BSA, 15 mM sodium azide, before the addition of glycerol to 40%

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CK15
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR