Anti-TGM2 Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-796EA 3590 DKK
PRSI49-796
Anti-TGM2 Rabbit Polyclonal Antibody
Antistoffer
Transglutaminases (TGases) are calcium-dependent enzymes catalysing the post-translational cross-linking of proteins.

Transglutaminase 2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

Protein A Column

PBS, pH 7.4, 0.2% BSA, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TGM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR