Anti-COL18A1 Mouse Monoclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-791EA 3840 DKK
PRSI49-791
Anti-COL18A1 Mouse Monoclonal Antibody
Antistoffer
Suitable for use in Immunohistochemistry and Immunoblot. Other applications not tested. Recognizes endostatin, Mr 22kD, an unidentified protein is also weakly detected, Mr 54kD. Species Crossreactivity: Human and mouse. Predicted to crossreact with rat.

COL18A1 antibody can be used in ELISA, and immunohistochemistry starting at 5 μg/mL.

Thiophilic Adsorption Chromatography

50 mM sodium phosphate, pH 8.0, 0.05% sodium azide, before the addition of glycerol to 30%

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: COL18A1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR