Anti-SAA1 Mouse Monoclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-782EA 3590 DKK
PRSI49-782
Anti-SAA1 Mouse Monoclonal Antibody
SAA1 antibody can be used in immunohistochemistry starting at 20 μg/mL.

Protein G Column

PBS, pH 7.2.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SAA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR