Anti-B2M Mouse Monoclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-779EA 0 DKK
PRSI49-779
Anti-B2M Mouse Monoclonal Antibody
Antistoffer
Beta 2-microglobulin is the low molecular weight component of HLA antigens and as such is involved in the presentation of endogenous antigens, activation of T lymphocytes and functional emergence of natural killer cells. It is found in the serum of normal individuals and in the urine in elevated amounts in patients with Wilson disease, cadmium poisoning and other conditions leading renal tubular dysfunction. ESTs have been isolated from libraries generated using virtually every tissue in the body.

B2M antibody can be used in immunohistochemistry starting at 20 μg/mL.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: B2M
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR