Anti-MMP11 Mouse Monoclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-774EA 3590 DKK
PRSI49-774
Anti-MMP11 Mouse Monoclonal Antibody
Antistoffer
MMP11 antibody can be used in ELISA, Western Blot starting at 1:250 - 1:1500, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, pH 7.4, 0.2% BSA, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MMP11
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR