Anti-ENG Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-773EA 3840 DKK
PRSI49-773
Anti-ENG Rabbit Polyclonal Antibody
Antistoffer
CD105 or Endoglin or GP160 is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists as an Oand N-glycosylated homodimer. Up-regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation-associated endothelial cell marker. CD105 binds to TGF-beta1 and Beta3 with high affinity but not TGF-beta2. Endoglin is a homodimeric transmembrane glycoprotein highly expressed by endothelial cells. It is a component of the transforming growth factor beta receptor complex as it binds TGFB1 and TGFB3 with high affinity. Endoglin has been shown to be a useful marker to identify tumor angiogenesis. Mutations in the endoglin gene produce hereditary hemorrhagic telangiectasia.

Endoglin antibody can be used in ELISA, and immunohistochemistry starting at 5 μg/mL.

10 mM PBS, pH 7.6, 0.2% BSA, 15 mM sodium azide, before the addition of glycerol to 40%

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ENG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR