Anti-COL18A1 Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-772EA 3840 DKK
PRSI49-772
Anti-COL18A1 Rabbit Polyclonal Antibody
Antistoffer
COL18A1 antibody can be used in ELISA, Western Blot starting at 1:2000, immunohistochemistry starting at 2.5 μg/mL, immunoprecipitation starting at 1:100, and flow cytometry.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: COL18A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR