Anti-SERPINH1 Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-769EA 3590 DKK
PRSI49-769
Anti-SERPINH1 Rabbit Polyclonal Antibody
CBP1 is a collagen binding protein located on endoplasmic reticulum. It shows sequence homology to the serine protease inhibitor serpin but does not bind endogenous proteases. CBP1 is involved in the intracellular processing, folding, assembly and secretion of procollagens.

SERPINH1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein A Column

PBS, 30% glycerol, 0.5% BSA, 0.01% Thimerosal.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SERPINH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR