Anti-S1PR1 Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-750EA 0 DKK
PRSI49-750
Anti-S1PR1 Rabbit Polyclonal Antibody
Antistoffer
EDG1, a Lysophospholipid/Lysosphingolipid Receptor, binds sphingosine-1-phosphate and may control cell proliferation, survival, and suppression of apoptosis. Overexpression of EDG-1 leads to increased cell-cell aggregation and overexpression of cadherins.

EDG1 antibody can be used in ELISA starting at 1:8000 - 1:32000, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: S1PR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR