Anti-XK Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-749EA 0 DKK
PRSI49-749
Anti-XK Rabbit Polyclonal Antibody
Antistoffer
XK antibody can be used in ELISA starting at 1:4000 - 1:20000, Western Blot starting at 1:250 - 1:2000, immunohistochemistry starting at 2.5 μg/mL, and immunofluorescence.

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: XK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR