Anti-MMP10 Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-703EA 0 DKK
PRSI49-703
Anti-MMP10 Rabbit Polyclonal Antibody
Antistoffer
MMP10 is a Metallopeptidase M10A type protease. Stromelysin2/MMP10 is a member of a gene family of metalloproteinases involved in extracellular matrix remodeling in normal and diseased processes. It degrades collagen and many non-collagenous components of the extracellular matrix including fibronectin, gelatins of type I, III, IV, and V and may play a role in the activation of latent procollagenase.

MMP10 antibody can be used in ELISA, immunofluorescence, and immunohistochemistry starting at 2.5 μg/mL.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MMP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR