Anti-PRKCA Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-690EA 3590 DKK
PRSI49-690
Anti-PRKCA Rabbit Polyclonal Antibody
Antistoffer
Protein Kinase C alpha antibody can be used in ELISA, immunohistochemistry starting at 1 - 2.5 μg/mL, and flow cytometry.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PRKCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR