Anti-PDGFRA Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-686EA 3590 DKK
PRSI49-686
Anti-PDGFRA Rabbit Polyclonal Antibody
Antistoffer
Platelet-derived Growth Factor Receptor Alpha (PDGFR alpha), a member of the a PDGF Receptor tyrosine kinase family, is a mediator of the biological actions of platelet-derived growth factor (PDGF). PDGF binding to PDGF receptors result in stimulation of cell growth, chemotaxis, and cell shape changes. Unlike PDGFR beta, PDGFR alpha can bind to both PDGF subunits. PDGFR alpha has been implicated in the development of spina bifida. In various mouse models, deregulated PDGFR alpha expression was found to result in congenital neural tube defects. In humans, various PDGFRA promoter haplotypes have been associated with neural tube defects.

PDGFRA antibody can be used in ELISA, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDGFRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR