Anti-MMP7 Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-684EA 0 DKK
PRSI49-684
Anti-MMP7 Rabbit Polyclonal Antibody
Antistoffer
MMP7 is a Metallopeptidase M10A type protease. MMP-7 (Matrilysin) is constitutively produced by exocrine epithelial cells through out the body. It is capable of degrading casein, fibronectin, gelatins of types 1, III, IV, and V, in addition to activating procollagenase. It has been shown to play a key role in invasion by carcinomas.

MMP7 antibody can be used in ELISA starting at 1:2000 - 1:8000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 2.5 μg/mL.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR