Anti-ITGB1BP1 Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-680EA 0 DKK
PRSI49-680
Anti-ITGB1BP1 Rabbit Polyclonal Antibody
Antistoffer
ITGB1BP1 antibody can be used in ELISA starting at 1:30000 - 1:90000, Western Blot starting at 1:400 - 1:2000, immunohistochemistry starting at 1:500, and immunofluorescence.

Protein G Column

PBS, 0.2% gelatin, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ITGB1BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR