Anti-DNMT3L Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-665EA 3590 DKK
PRSI49-665
Anti-DNMT3L Rabbit Polyclonal Antibody
Antistoffer
DNA cytosine-5-methyltransferase-3 proteins are involved in de novo methylation of CpG islands. Mutations in the DNMT3B gene cause the ICF immunodeficiency syndrome. DNA methyltransferases and regulatory factors are likely to be involved in the establishment of maternal imprints in growing diplotene oocytes and paternal imprints in perinatal prospermatogonia. Imprinting is essential for viable development. Expression of the DNMT3L mRNA was not detectable by Northern blotting; however, RT-PCR amplification revealed that it is expressed at low levels in several tissues including testis, ovary, and thymus.

DNMT3L antibody can be used in ELISA, Western Blot starting at 1:1000, immunohistochemistry starting at 1.25 μg/mL, immunocytochemistry, and immunoprecipitation.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DNMT3L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR