Anti-DNMT1 Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-664EA 3590 DKK
PRSI49-664
Anti-DNMT1 Rabbit Polyclonal Antibody
Antistoffer
DNA (cytosine-5-)-methyltransferase 1 has a role in the establishment and regulation of tissue-specific patterns of methylated cytosine residues. Aberrant methylation patterns are associated with certain human tumors and developmental abnormalities.

DNMT1 antibody can be used in ELISA starting at 1:5000 - 1:10000, Western Blot starting at 1:5000 - 1:10000, immunohistochemistry starting at 1.25 μg/mL, and immunoprecipitation starting at 1:100

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DNMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR