Anti-BMP3 Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-647EA 0 DKK
PRSI49-647
Anti-BMP3 Rabbit Polyclonal Antibody
BMP3 belongs to the transforming growth factor-beta (TGFB) superfamily. Bone morphogenic protein induces bone formation.

BMP3 antibody can be used in Western Blot, immunohistochemistry starting at 2.5 mg/ml, and flow cytometry starting at 1/50 - 1/100.

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR