Anti-BMP1 Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-645EA 0 DKK
PRSI49-645
Anti-BMP1 Rabbit Polyclonal Antibody
Antistoffer
BMP1 encodes a Metallopeptidase M12A type protease that is comprised of an amino terminal signal peptide, prodomain and an astacin-like protease domain followed by CUB and EGF-like motifs. BMP1 protein has been shown to cleave chordin, the C-terminal propeptides of procollagen I, II, III and VII, pro-lysyl oxidase, probiglycan, and laminin-5, and its expression has been shown to induce cartilage and bone formation in vivo. At least six spliced variants have been reported. These share the N-terminal protease domain but differ in the C-terminus, and thus contain varying numbers of CUB and EGF-like domains.

BMP1 antibody can be used in ELISA starting at 1:10000 - 1:50000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 2.5 μg/mL, and immunofluorescence.

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR