Anti-BMP2K Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-644EA 3590 DKK
PRSI49-644
Anti-BMP2K Rabbit Polyclonal Antibody
Antistoffer
BIKE is a NAK serine/threonine protein kinase that mediates osteoblast differentiation. The protein contains a N-terminal serine/threonine kinase domain, a C-terminal bipartite nuclear localization signal and a glutamine-rich region that may function in protein-protein interactions. At least two mRNA transcript variants, resulting from alternative splicing at the BIKE locus, have been reported.

BMP2K antibody can be used in ELISA starting at 1:20000 - 1:80000, Western Blot starting at 1:1000 - 1:3000, immunohistochemistry starting at 2.5 μg/mL, and immunofluorescence.

Protein G Column

PBS, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BMP2K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR