Anti-PPP4C Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-620EA 3590 DKK
PRSI49-620
Anti-PPP4C Rabbit Polyclonal Antibody
Protein phosphatase 4 (protein phosphatase X) is a 35kD serine/threonine phosphatase with predicted binding sites for iron and manganese. It is found in both the cytoplasm and the nucleus. The phophoprotein alpha 4 has been identified as a potential regulatory subunit. PPP4C has been shown to localize to centrosomes/spindle pole bodies and is believed to be involved in microtubule organization. It may also play a role in relaying the TNF-alpha signal to the JNK pathway. Additionally, it has been shown to associate with C-Rel, NF-Kappa B p50, and RelA. Hence, it may be involved in the activation of these key mediators of immune response and apoptosis.

Protein Phosphatase 4 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Purified

PBS, 0.08% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PPP4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR