Anti-PPP2R1A Rabbit Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-619EA 0 DKK
PRSI49-619
Anti-PPP2R1A Rabbit Polyclonal Antibody
PPP2R1B antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Ammonium Sulfate Precipitation

PBS, 0.08% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PPP2R1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR