Anti-IKBKB Chicken Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-609EA 3960 DKK
PRSI49-609
Anti-IKBKB Chicken Polyclonal Antibody
Antistoffer
IKKbeta, an IKAPPAB-type protein kinase, is a member of the multimeric IkappaB kinase complex. This complex, often consisting of IKKalpha/IKKbeta heterodimers plus regulatory IKKgamma subunits, phosphorylates IkappaB proteins, marking them for destruction and thereby allowing activation of NFkappaB and downstream nuclear genes. The constitutively expressed IKKbeta protein contains an N-terminal kinase domain, a central leucine zipper motif, and a C-terminal helix-loop-helix motif. In knockout studies, IKKbeta (-/-) mice died between embryonic day 12.5 and 14 due to extensive liver damage from apoptosis; however, they were rescued by inactivation of the tumor necrosis factor receptor 1 gene. IKKbeta has been implicated in the pathogenesis of insulin resistance in obesity and type II diabetes mellitus.

IKBKB antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunoprecipitation

PBS. No preservative added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IKBKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR