Anti-DNMT1 Chicken Polyclonal Antibody

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI49-604EA 3960 DKK
PRSI49-604
Anti-DNMT1 Chicken Polyclonal Antibody
DNA (cytosine-5-)-methyltransferase 1 has a role in the establishment and regulation of tissue-specific patterns of methylated cytosine residues. Aberrant methylation patterns are associated with certain human tumors and developmental abnormalities.

DNMT1 antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1 - 5 μg/mL, immunohistochemistry starting at 5 - 10 μg/mL, immunohistochemistry in frozen tissues starting at 1 - 10 μg/mL, and flow cytometry.

PBS. No preservative added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DNMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR