Keep my session open?
Ending In 
The session is expired
Din session er udløbet. For din sikkerhed har vi logget dig ud.
Vil du logge ind igen?
OBS! Det er ikke længere muligt at skrive til @dk.vwr.com benyt istedet @vwr.com. Kontakt evt vores kundeservice.

Anti-TRAF2 And NCK Interacting Kinase Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
TNIK is a MSN protein kinase that interacts with both TNF receptor-associated factor 2 (TRAF2) and the adapter protein NCK. The protein has been shown to activate the c-Jun N-terminal kinase pathway when overexpressed in Phoenix-A cells, and this activation is mediated solely by the C-terminal germinal center kinase homology region. TNIK has been shown to phosphorylate gelsolin, the principal intracellular and extracellular actin-severing protein, in vitro. This and evidence from mutational studies suggest that TNIK functions in the regulation of the cytoskeleton. At least eight splice isoforms have been reported.

TNIK antibody can be used in immunohistochemistry starting at 30 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRAF2 And NCK Interacting Kinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey




 

BESTIL

Udvid alle / Skjul alle
Størrelse Pakn VWR katalog nummer Mængde
50 µg 50 µG PRSI48-331
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Specifikationer

Antigen symbol TNIK
Antigen name Traf2 And Nck Interacting Kinase
Reactivity Gorilla, Gibbon, Human, Monkey
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
ELISA Yes
ImmunoChemistry Yes
Immunogen TNIK antibody was raised against a peptide located near the internal domain of TNIK (Human).
Form Liquid
Tested applications Immunoaffinity Chromatography

Skriv en bedømmelse

Brug for hjælp?

  • Uanset hvad du mener, positivt eller negativt, er det vigtigt for os.
  • Det tager kun få sekunder at give os feedback.
  • Din feedback vil blive offentliggjort på internettet.

VWR.com vil offentliggøre dit navn som vwr.com bruger.
Du kan bruge dette navn, eller redigere og gemme et kaldenavn.
Vi anbefaler at brugerne bruger deres første og sidste initialer

Bekræft venligst branchen du arbejder i

Eksempel: farmaceutisk

Bekræft venligst dit arbejde

Eksempel: Klinisk assistent

Bekræft venligst Byen

Eksempel: New York

Bedømmelser

Samlet vurdering
Gennemsntilig vurdering

Sortér alle efter:

Enter Details

Select the article for which you are looking for Certificate

Anfør Lotnummeret. Feltet er mandatory. skriv "x" for VWR kemikalier hvis du ikke kender nummeret

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivelse katalog Nummer Availability Enhed Din pris Price Per Antal