Anti-GRM2 Rabbit Polyclonal Antibody
Leverandør: ProSci
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PRSI48-303EA 3590 DKK
PRSI48-303
Anti-GRM2 Rabbit Polyclonal Antibody
The Metabotropic Glutamate Receptor GRM2 binds L-glutamate and has been linked to the inhibition of the cyclic AMP signaling cascade. GRM2 is involved in mediating the suppression of neurotransmission and in synaptogenesis and synaptic stabilization. Glutamatergic neurotransmission plays a role in most aspects of normal brain function and can be perturbed in many neuropathologic conditions.
GRM2 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunoprecipitation
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GRM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Chimpanzee, Gorilla, Human, Monkey, Rabbit