VWR®, Bags, Blue Twist-Seal
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
129-9608EA 4340 DKK
129-9608 129-9609 129-9614
VWR®, Bags, Blue Twist-Seal
Poser Prøveposer
This line of blue twist-seal printed bags, with safety tab and two round wires, has been specifically designed for food industry. The bags accommodate solid, semi-solid and liquid compounds.
  • Blue tear-off for increased visibility
  • Distinct colour avoids possible foreign matter contamination
  • Visual recognition of the product, but can also be used to segregate within a factory environment
  • Made from virgin PE
  • Optimised opening area to facilitate insertion of sample
  • Lot number traceability
  • Bags accommodate solid, semi-solid and liquid compounds
  • All sterile sampling bags are also free from RNase, DNase and pyrogens
Certificeringer: Food contact approved.