Du Søgte efter: Septa
80  results were found
SearchResultCount:"80"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: Merck
Beskrivelse: Ready to use GC septa for inlet temperature of up to 100 to 350 °C.
Leverandør: Merck
Beskrivelse: These septa are suitable for use with 50 to 300 °C inlet temperatures.
Leverandør: VWR Collection
Beskrivelse: Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
Leverandør: VWR Collection
Beskrivelse: Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
Leverandør: VWR Collection
Beskrivelse: Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
Leverandør: VWR Collection
Beskrivelse: Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
Leverandør: VWR Collection
Beskrivelse: Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
Leverandør: VWR Collection
Beskrivelse: Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
Leverandør: VWR Collection
Beskrivelse: VWR®, Septa, Beige silicone/white PTFE, Hårdhed: 50° shore A, Tykkelse: 3,2 mm, diameter: 43 mm
Varenummer: (548-2358)
Leverandør: VWR Collection
Beskrivelse: VWR®, Septa, Silicone blue transparent/PTFE white, Hårdhed: 45° shore A, Tykkelse: 3,0 mm
UOM: 1 * 1.000 Items
Varenummer: (548-0574)
Leverandør: VWR Collection
Beskrivelse: VWR®, Septa, Natural rubber red-orange/TEF transparent, Hårdhed: 60° shore A, Tykkelse: 1,3 mm
UOM: 1 * 1.000 Items
Leverandør: VWR Collection
Beskrivelse: Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
Leverandør: VWR Collection
Beskrivelse: Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
Leverandør: VWR Collection
Beskrivelse: Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
Varenummer: (548-0654A)
Leverandør: VWR Collection
Beskrivelse: VWR®, Septa, Cream silicone/beige PTFE, Hårdhed: 50° shore A, Tykkelse: 3,2 mm, diameter: 30 mm
UOM: 1 * 100 Items
Leverandør: VWR Collection
Beskrivelse: Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 80
no targeter for Bottom