J.T.Baker® HYDRA-POINT™, Coulometric Reagents for Water Determination by Karl Fischer Titration
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J.T. Baker® HYDRA-POINT™
6287-01EA 1620 DKK
6287-01 6280-01 6285-01 6287-02 6280-02 6285-02 6281-00 6284-01 6284-02 6283-01
J.T.Baker® HYDRA-POINT™, Coulometric Reagents for Water Determination by Karl Fischer Titration
Karl Fischer titranter og reagenser
HYDRA-POINT coulometric reagents are designed to deliver faster analyses, lower detection limits and increased accuracy.
  • Pyridine- and chlorocarbon-free vessel solution
  • The fritless/diaphragmless reagent works both in cells without and with diaphragm
  • High water capacity, where the anolyte can take over 1000 mg of water per charge
These products feature lower background noise that gives a better signal/noise ratio, resulting in a significant increase in the lower detection limit—to as low as 1 ppm.