Vacuum filtration manifolds
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
516-7026EA 0 DKK
SART16828
Vacuum filtration manifolds
These traditional multi-branch stainless steel manifold systems are available with 100 ml or 500 ml capacity funnels. The three or six separate filter holders save time when mass examinations have to be carried out. Due to the stainless steel taps on the manifold ports, the vacuum for each holder can be turned on and off individually.
  • The stainless steel frit allows homogenous distribution of the residues on the membrane filter surface
  • Funnel and filter support can be disinfected by flaming
Filtration area: 12,5 cm²
For filter diameter: 47 or 50 mm
Max. operating pressure: Vacuum or max. 2 bar pressure