Natriumfluorid 98.0-102.0% (dry basis), pulver USP, J.T.Baker®

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

3689-09EA 19960 DKK
3689-09 3689-08 3689-R
Natriumfluorid 98.0-102.0% (dry basis), pulver USP, J.T.Baker®
Sodium fluoride
Formel: NaF
Molekylvægt: 41,99 g/mol
Kogepunkt: 1700 °C (1013 hPa)
Smeltepunkt: 993 °C
Massefylde: 2,8 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003524
CAS-nummer: 7681-49-4
EINECS: 231-667-8
UN nummer: 1690
ADR: 6.1,III
Merck Index: 12,08762
https://dk.vwr.com/stibo/web/std.lang.all/06/30/16640630.jpg

Bestil nu

Specifikationer Testresultater

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acidity or Alkalinity Passes Test
USP - Loss on Drying ≤1.0 %
USP - Fluosilicate (volume of 0.10N NaOH required) ≤1.5 ml
USP - Chloride (Cl) ≤0.012 %
USP - Assay (NaF) (dried basis) 98.0 - 102.0 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR