Wipe rolls for impregnation, with dispensing bucket
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
115-2420EA 368 DKK
VWRI115-2420
Wipe rolls for impregnation, with dispensing bucket
These strong centre-feed wipes, made from 70% viscose and 30% polyester, are supplied with a resealable dispensing bucket for custom impregnation.
  • High absorption capacity
  • For pre-impregnation at the point of use
  • Can be impregnated with any solution