VWR®, Kitchen Wipes, HACCP
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-2413EA 588 DKK
115-2413 115-2404
VWR®, Kitchen Wipes, HACCP
Papirservietter Genanvendelige rengøringsklude Klude til alment brug
These reusable wipes are made from 55% viscose, 35% polyester and 10% binder. Available in four different colours.
  • Can be rinsed and reused
  • High absorption
  • Non scratching
  • Recyclable
  • Food contact certified
  • HACCP colour coding
Typical applications include: Food and beverage, healthcare and janitorial tasks.
Basis weight: 45 g/m²
Absorption: ≥500%