Surgical mask, tie-on, type II
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ANSM85002EA 46.9 DKK
ANSM85002
Surgical mask, tie-on, type II
Masker Kirurgmasker
These masks help prevent large particles expelled by the wearer such as spit or mucous from reaching the patient or work environment.
  • Pleat style with ties
  • Protective three-layer construction
Appropriate for situations where exposure to blood and other bodily fluids from the patient or work environment.