Industrial wipes, DRC63, VWR®
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-2393EA 735 DKK
VWRI115-2393
Industrial wipes, DRC63, VWR®
These wet-laid, double re-creped, general purpose wipes, made from wood pulp with binder, can be used for many different appplications.
  • Excellent absorption capacity for water and oil
  • Resistant and non scratching
  • Stronger and more absorbent than standard kitchen roll
  • Recyclable
  • Food contact certified
Typical applications include: Food and beverage, automotive, industry, printing, healthcare and janitorial tasks.
Basis weight: 63 g/m²
Absorption: 760%