VWR®, Disposable Particle Filtering Respirators, FFP3
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
111-2279EA 270 DKK
111-2279
VWR®, Disposable Particle Filtering Respirators, FFP3
Åndedrætsværn Engangsmasker Partikelmasker
Disposable flat-fold three layer P3 respirator; spunbonded PP outer layer, meltblown PP filter media and spunbonded PP inner layer.
  • Low breathing resistance
  • Twin elastic headbands
  • Adjustable aluminium nose bridge
  • Comfortable foam seal
Compatible with eyewear. For use in lab, cleanroom production or research.
Certificeringer: EN 149:2001
Emballering: Individual hygienically packed.
Advarsel: * One size fits all.