Reusable earplugs, 1291, E-A-R™ Ultrafit™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E-A-R™ UltraFit™
111-1130EA 1940 DKK
111-1130 111-0450
Reusable earplugs, 1291, E-A-R™ Ultrafit™
Ørepropper Genanvendelige ørepropper
These earplugs are made from flexible materials that are already pre-formed to ensure optimum fit.
  • The triple-flange feature of the earplugs ensures a perfect fit: No size adjustment required
  • Easy to insert
  • Washable and reusable
SNR: 32 dB
Certificeringer: EN 352-2
Advarsel: * One size fits all.