MXP high pressure injection valve, Rheodyne®, MX Series II™
Leverandør: IDEX Health & Science
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
554-3290EA 33160 DKK
RHEOMXP7920-000
MXP high pressure injection valve, Rheodyne®, MX Series II™
This high pressure actuated valve is designed to be added to an existing instrument or to work optimally with stand-alone lab configurations.
  • Can withstand up to 6000 psi (414 bar)
  • Chemically compatible for use with most mobile phase compositions
  • Available with analytical and nano-scale flow paths
  • Multiple control options
  • Stand alone capability
  • Included software