Disposable earplugs, E-A-R™ Classic™ II
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E-A-R™ Classic™
111-1102EA 0 DKK
111-1102
Disposable earplugs, E-A-R™ Classic™ II
Ørepropper Ørepropper til engangsbrug
E-A-R Classic II earplugs are made from soft, energy-absorbing polymer foam that effectively seals off the ear canal and ensures all-day comfort.
  • Adapt to fit most ear canals
  • Moisture-proof, prevents slippage due to perspiration
  • Roughened surface prevents slippage and ensures reliable protection
  • Dermatologically safe foam is non irritating
SNR: 28 dB
Certificeringer: EN 352-2
Advarsel: * One size fits all.