Anti-CHD3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BOSSBS-8162R-A555EA 6410 DKK
BOSSBS-8162R-A555
Anti-CHD3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Component of the histone deacetylase NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Required for anchoring centrosomal pericentrin in both interphase and mitosis, for spindle organization and centrosome integrity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CHD3
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR