Anti-RPL15 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BOSSBS-6571R-A555EA 6380 DKK
BOSSBS-6571R-A555
Anti-RPL15 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L15E family of ribosomal proteins. It is located in the cytoplasm. This gene shares sequence similarity with the yeast ribosomal protein YL10 gene. Although this gene has been referred to as RPL10, its official symbol is RPL15. This gene has been shown to be overexpressed in some esophageal tumors compared to normal matched tissues. Alternate splicing results in multiple transcript variants. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RPL15
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR