Anti-IL18BP Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BOSSBS-4040R-CY7EA 6470 DKK
BOSSBS-4040R-CY7
Anti-IL18BP Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Isoform A binds to IL-18 and inhibits its activity. Functions as an inhibitor of the early TH1 cytokine response.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL18BP
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR