Keep my session open?
Ending In 
Din session er udløbet.
Din session er udløbet. For din sikkerhed har vi logget dig ud.
Vil du logge ind igen?
OBS! @vwr.com benyttes ikke længere – benyt i stedet @avantorsciences.com. Kontakt evt vores kundeservice.
Water purification systems, TU+
Leverandør: VWR Collection
Bedømmelser
Samlet vurdering
Gennemsntilig vurdering
Sortér alle efter:
171-1239EA 48310 DKK InStock
171-1239 171-1237 171-1236 171-1231 171-1233 171-1232 171-1234 171-1238 171-1235
Water purification systems, TU+
Vandrensningssystemer Rene og ultrarene vandsystemer
VWR water purification systems TU 3+, 6+ and 12+ ultrapure systems produce up to 100 litres/day of Type 1 ultrapure water from tap water, as well as Type 2 water available from the tank. The TU 3+ and 6+ feature a built-in 6 litre sterile vented reservoir to store Type 2 water. The TU 12+ offers the choice of a 30 or 60 litre external reservoir for customisable storage capacity. Each system is available in three different models: Basic, with UV lamp (UV) or UV lamp and ultrafilter (UV/UF) to fullfil the specific demand of each application. Typical applications include: molecular biology and microbiology (cell and tissue culture, PCR, DNA sequencing, electrophoresis) and analytical chemistry (HPLC, GC, GC-MS, ICP-MS, AA, TOC measurements, routine laboratory work...).
  • Easy to install and maintain
  • Elegant symplicity, perfomance and value
  • Dispensing is easy and features variable speed to control flow
  • The display can be tilted for optimal reading
  • Placement options: Can be placed on a laboratory bench or mounted on the wall
  • Conical bottom outlet allows for complete draining and efficient cleaning plus disinfection
  • PE reservoir is opaque to light; CO2 adsorber prevents TOC value increase from drawn-in CO2
  • Optional UV lamp and ultrafilter to customise ultrapure water quality
  • Sterile venting filter/reservoir overflow protects against contamination by microorganisms
  • Recirculation pump protects the high purity water from bacterial growth during standstills and maintains low conductivity value
  • Exchanging pre-treatment cartridges is easy without any system depressurisation
Water purification progresses through independent cartridges with Aquastop quick-connect for fast replacement.
- Module 1 - Combination of pre-treatment and reverse osmosis membrane
- Module 2 - Polishing cartridge contains high quality ultrapure resin for consistent purity and long cartridge life
A pre-treatment filter may be required if the feed water has high levels of free chlorine or hardness as this can damage the cartridge. High levels of particles and colloids may also block the reverse osmosis membrane. Please contact VWR for the right solution.
Each model uses an internal recirculation mode to ensure availability of high purity water at any time.
Leveringsinformation: TU 3+, 6+ and 12+ systems include an RO/pre-treatment cartridge, polisher cartridge pack, 0,2 µM final filter, optional UV lamp and ultrafilter.
Order Now
BESTIL
Specifikationer
TILBEHØR
RELATEREDE OPLYSNINGER
Skriv en bedømmelse
Bedømmelser
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse katalog Nummer Availability Enhed Din pris Price Per Antal